cis-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate

Lieferant: Apollo Scientific

OR307699-1G OR307699-250MG
APOSOR307699-1GEA 509 EUR
APOSOR307699-1G APOSOR307699-250MG
cis-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
cis-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
Formel: C₁₃H₁₆BrNO₂
Molecular Weight: 298.176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24368283
CAS-Nummer: 1431365-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR