trans-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
Lieferant: Apollo Scientific
OR307700-1G
OR307700-250MG
APOSOR307700-1GEA
509
EUR
APOSOR307700-1G
APOSOR307700-250MG
trans-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
trans-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
Formel:
C₁₃H₁₆ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD24368284 CAS-Nummer: 1416307-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...