cis-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate

Lieferant: Apollo Scientific

OR307706-1G OR307706-250MG
APOSOR307706-1GEA 509 EUR
APOSOR307706-1G APOSOR307706-250MG
cis-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate
cis-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate
Formel: C₁₃H₁₆BrNO₂
Molecular Weight: 298.176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26936317
CAS-Nummer: 1980008-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR