(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate

Lieferant: Apollo Scientific

Synonyme: (-)-(5aS,10bR)-5a,10b-Dihydro-2-mesityl-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium Chloride Monohydrate, Bode Catalyst 1, (-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium Chloride Monohydrate, Bode Catalyst 1 Monohydrate

OR307708-250MG
APOSOR307708-250MGEA 325 EUR
APOSOR307708-250MG
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate
Formel: C₂₁H₂₂ClN₃O
Molecular Weight: 367,88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566990
CAS-Nummer: 919102-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR