4-(3-Methoxy-benzyl)-piperidine
Lieferant: Apollo Scientific
OR307711-1G
APOSOR307711-1GEA
499
EUR
APOSOR307711-1G
4-(3-Methoxy-benzyl)-piperidine
4-(3-Methoxy-benzyl)-piperidine
Formel:
C₁₃H₁₉NO Molecular Weight: 205,296 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863572 CAS-Nummer: 150019-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...