4-(2-Methoxy-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307712-1G
APOSOR307712-1GEA 441 EUR
APOSOR307712-1G
4-(2-Methoxy-benzyl)-piperidine
4-(2-Methoxy-benzyl)-piperidine
Formel: C₁₃H₁₉NO
Molecular Weight: 205,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863573
CAS-Nummer: 37581-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR