4-(2-Methoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-(2-Methoxybenzyl)piperidine hydrochloride

OR307713-1G
APOSOR307713-1GEA 174 EUR
APOSOR307713-1G
4-(2-Methoxy-benzyl)-piperidine hydrochloride
4-(2-Methoxy-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₉NO·ClH
Molecular Weight: 241,757 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437508
CAS-Nummer: 37581-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR