4-(4-Ethoxybenzyl)piperidine
Lieferant: Apollo Scientific
OR307714-250MG
APOSOR307714-250MGEA
219
EUR
APOSOR307714-250MG
4-(4-Ethoxybenzyl)piperidine
4-(4-Ethoxybenzyl)piperidine
Formel:
C₁₄H₂₁NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD05189928 CAS-Nummer: 893754-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...