4-(3-Ethoxy-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307715-1G
APOSOR307715-1GEA 499 EUR
APOSOR307715-1G
4-(3-Ethoxy-benzyl)-piperidine
4-(3-Ethoxy-benzyl)-piperidine
Formel: C₁₄H₂₁NO
Molecular Weight: 219,323 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863574
CAS-Nummer: 782504-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR