4-(3-Ethoxy-benzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
OR307716-1G
APOSOR307716-1GEA
433
EUR
APOSOR307716-1G
4-(3-Ethoxy-benzyl)-piperidine hydrochloride
4-(3-Ethoxy-benzyl)-piperidine hydrochloride
Formel:
C₁₄H₂₁NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD10703294 CAS-Nummer: 1172021-56-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...