4-(4-Methyl-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307717-1G
APOSOR307717-1GEA 375 EUR
APOSOR307717-1G
4-(4-Methyl-benzyl)-piperidine
4-(4-Methyl-benzyl)-piperidine
Formel: C₁₃H₁₉N
Molecular Weight: 189,297 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05189925
CAS-Nummer: 92822-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR