4-(4-Ethyl-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307719-1G
APOSOR307719-1GEA 467 EUR
APOSOR307719-1G
4-(4-Ethyl-benzyl)-piperidine
4-(4-Ethyl-benzyl)-piperidine
Formel: C₁₄H₂₁N
Molecular Weight: 203,323 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863575
CAS-Nummer: 781595-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR