2,2'-Bipyridine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR30772-1G OR30772-250MG OR30772-500MG
APOSOR30772-1GEA 700 EUR
APOSOR30772-1G APOSOR30772-250MG APOSOR30772-500MG
2,2'-Bipyridine-3-carboxylic acid
2,2'-Bipyridine-3-carboxylic acid
Formel: C₁₁H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11100052
CAS-Nummer: 220340-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR