2,2'-Bipyridine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR30772-1G
OR30772-250MG
OR30772-500MG
APOSOR30772-1GEA
700
EUR
APOSOR30772-1G
APOSOR30772-250MG
APOSOR30772-500MG
2,2'-Bipyridine-3-carboxylic acid
2,2'-Bipyridine-3-carboxylic acid
Formel:
C₁₁H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100052 CAS-Nummer: 220340-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...