4-Piperidin-4-ylmethyl-phenol hydrochloride

Lieferant: Apollo Scientific
Achtung

OR307720-1G
APOSOR307720-1GEA 491 EUR
APOSOR307720-1G
4-Piperidin-4-ylmethyl-phenol hydrochloride
4-Piperidin-4-ylmethyl-phenol hydrochloride
Formel: C₁₂H₁₇NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08437509
CAS-Nummer: 1171849-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR