4-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307722-1G
APOSOR307722-1GEA 441 EUR
APOSOR307722-1G
4-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride
4-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride
Formel: C₁₄H₂₁NO₂·ClH
Molecular Weight: 271,783 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703300
CAS-Nummer: 1172908-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR