4-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307724-1G
APOSOR307724-1GEA 467 EUR
APOSOR307724-1G
4-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine
4-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine
Formel: C₁₅H₂₃NO
Molecular Weight: 233,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863589
CAS-Nummer: 955314-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR