4-(4-Phenoxybenzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
OR307725-1G
APOSOR307725-1GEA
252
EUR
APOSOR307725-1G
4-(4-Phenoxybenzyl)-piperidine hydrochloride
4-(4-Phenoxybenzyl)-piperidine hydrochloride
Formel:
C₁₈H₂₁NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669676 CAS-Nummer: 1172749-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...