3-(4-Methoxy-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307726-1G
APOSOR307726-1GEA 467 EUR
APOSOR307726-1G
3-(4-Methoxy-benzyl)-piperidine
3-(4-Methoxy-benzyl)-piperidine
Formel: C₁₃H₁₉NO
Molecular Weight: 205,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863592
CAS-Nummer: 136422-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR