3-(3-Methoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307727-1G
APOSOR307727-1GEA 267 EUR
APOSOR307727-1G
3-(3-Methoxy-benzyl)-piperidine hydrochloride
3-(3-Methoxy-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₉NO·ClH
Molecular Weight: 241,757 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787524
CAS-Nummer: 179480-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR