3-(4-Ethoxy-benzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
OR307729-1G
APOSOR307729-1GEA
185
EUR
APOSOR307729-1G
3-(4-Ethoxy-benzyl)-piperidine hydrochloride
3-(4-Ethoxy-benzyl)-piperidine hydrochloride
Formel:
C₁₄H₂₁NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437511 CAS-Nummer: 1170629-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...