3-(3-Methyl-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307730-1G
APOSOR307730-1GEA 467 EUR
APOSOR307730-1G
3-(3-Methyl-benzyl)-piperidine
3-(3-Methyl-benzyl)-piperidine
Formel: C₁₃H₁₉N
Molecular Weight: 189,297 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863598
CAS-Nummer: 955287-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR