3-(2-Methylbenzyl)piperidine
Lieferant: Apollo Scientific
OR307731-1G
APOSOR307731-1GEA
509
EUR
APOSOR307731-1G
3-(2-Methylbenzyl)piperidine
3-(2-Methylbenzyl)piperidine
Formel:
C₁₃H₁₉N Molecular Weight: 189,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863599 CAS-Nummer: 955314-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...