4-Piperidin-3-ylmethyl-phenol hydrochloride

Lieferant: Apollo Scientific

OR307733-1G
APOSOR307733-1GEA 267 EUR
APOSOR307733-1G
4-Piperidin-3-ylmethyl-phenol hydrochloride
4-Piperidin-3-ylmethyl-phenol hydrochloride
Formel: C₁₂H₁₇NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669680
CAS-Nummer: 1170102-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR