3-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307734-1G
APOSOR307734-1GEA 161 EUR
APOSOR307734-1G
3-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride
3-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride
Formel: C₁₄H₂₁NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669681
CAS-Nummer: 625454-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR