3-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307736-1G
APOSOR307736-1GEA 467 EUR
APOSOR307736-1G
3-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine
3-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine
Formel: C₁₅H₂₃NO
Molecular Weight: 233.349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863615
CAS-Nummer: 779323-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR