3-(4-Phenoxy-benzyl)-piperidine hydrochloride
Lieferant: Apollo Scientific
OR307737-1G
APOSOR307737-1GEA
433
EUR
APOSOR307737-1G
3-(4-Phenoxy-benzyl)-piperidine hydrochloride
3-(4-Phenoxy-benzyl)-piperidine hydrochloride
Formel:
C₁₈H₂₁NO·ClH Molecular Weight: 303,826 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669689 CAS-Nummer: 1171985-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...