2-(3-Methoxy-benzyl)-piperidine

Lieferant: Apollo Scientific
Achtung

OR307738-1G
APOSOR307738-1GEA 499 EUR
APOSOR307738-1G
2-(3-Methoxy-benzyl)-piperidine
2-(3-Methoxy-benzyl)-piperidine
Formel: C₁₃H₁₉NO
Molecular Weight: 205.296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663630
CAS-Nummer: 108958-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR