2-(3-Ethoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307739-1G
APOSOR307739-1GEA 185 EUR
APOSOR307739-1G
2-(3-Ethoxy-benzyl)-piperidine hydrochloride
2-(3-Ethoxy-benzyl)-piperidine hydrochloride
Formel: C₁₄H₂₁NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669727
CAS-Nummer: 1170163-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR