6-Oxo-1,6-dihydropyridazine-4-carboxylic acid 95+%
Lieferant: Apollo Scientific
Synonyme:
1,6-Dihydro-6-oxopyridazine-4-carboxylic acid
OR30774-1G
APOSOR30774-1GEA
82.8
EUR
APOSOR30774-1G
6-Oxo-1,6-dihydropyridazine-4-carboxylic acid 95+%
6-Oxo-1,6-dihydropyridazine-4-carboxylic acid
Formel:
C₅H₄N₂O₃ Molecular Weight: 140,0969 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437486 CAS-Nummer: 867130-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...