2-(4-Methyl-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307740-1G
APOSOR307740-1GEA 0 EUR
APOSOR307740-1G
2-(4-Methyl-benzyl)-piperidine hydrochloride
2-(4-Methyl-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₉N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669734
CAS-Nummer: 782504-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR