2-(3-Methyl-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307741-1G
APOSOR307741-1GEA 0 EUR
APOSOR307741-1G
2-(3-Methyl-benzyl)-piperidine hydrochloride
2-(3-Methyl-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₉N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669735
CAS-Nummer: 1171524-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR