2-(2-Methoxy-benzyl)-piperidine

Lieferant: Apollo Scientific

OR307742-1G
APOSOR307742-1GEA 467 EUR
APOSOR307742-1G
2-(2-Methoxy-benzyl)-piperidine
2-(2-Methoxy-benzyl)-piperidine
Formel: C₁₃H₁₉NO
Molecular Weight: 205,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663626
CAS-Nummer: 383128-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR