2-(4-Methoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307743-1G
APOSOR307743-1GEA 433 EUR
APOSOR307743-1G
2-(4-Methoxy-benzyl)-piperidine hydrochloride
2-(4-Methoxy-benzyl)-piperidine hydrochloride
Formel: C₁₃H₁₉NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08669737
CAS-Nummer: 420137-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR