2-(4-Ethoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307744-1G
APOSOR307744-1GEA 433 EUR
APOSOR307744-1G
2-(4-Ethoxy-benzyl)-piperidine hydrochloride
2-(4-Ethoxy-benzyl)-piperidine hydrochloride
Formel: C₁₄H₂₁NO·ClH
Molecular Weight: 255,784 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669738
CAS-Nummer: 1172745-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR