2-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307745-1G
APOSOR307745-1GEA 433 EUR
APOSOR307745-1G
2-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine hydrochloride
2-(4-Methoxy-3,5-dimethyl-benzyl)-piperidine hydrochloride
Formel: C₁₅H₂₃NO·ClH
Molecular Weight: 269.81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669739
CAS-Nummer: 1172932-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR