2-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride

Lieferant: Apollo Scientific

OR307746-1G
APOSOR307746-1GEA 433 EUR
APOSOR307746-1G
2-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride
2-(3,4-Dimethoxy-benzyl)-piperidine hydrochloride
Formel: C₁₄H₂₁NO₂·ClH
Molecular Weight: 271,783 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669740
CAS-Nummer: 109247-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR