1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

OR307749-1G OR307749-500MG
APOSOR307749-1GEA 0 EUR
APOSOR307749-1G APOSOR307749-500MG
1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic acid
1-(3,4-Dimethoxyphenyl)cyclopropanecarboxylic acid
Formel: C₁₂H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08442257
CAS-Nummer: 862821-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR