1-(3-Bromphenyl)cyclopropancarbonitril

Lieferant: Apollo Scientific

OR307750-1G
APOSOR307750-1GEA 126 EUR
APOSOR307750-1G
1-(3-Bromphenyl)cyclopropancarbonitril
1-(3-Bromphenyl)cyclopropancarbonitril
Formel: C₁₀H₈BrN
Molecular Weight: 222,0812 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374414
CAS-Nummer: 124276-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR