1-(3,4-Dimethoxyphenyl)cyclopropanecarbonitrile

Lieferant: Apollo Scientific

OR307752-1G
APOSOR307752-1GEA 267 EUR
APOSOR307752-1G
1-(3,4-Dimethoxyphenyl)cyclopropanecarbonitrile
1-(3,4-Dimethoxyphenyl)cyclopropanecarbonitrile
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314306
CAS-Nummer: 20802-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR