(1-(3-Bromophenyl)cyclopropyl)methanamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR307754-1G
APOSOR307754-1GEA
401
EUR
APOSOR307754-1G
(1-(3-Bromophenyl)cyclopropyl)methanamine hydrochloride
(1-(3-Bromophenyl)cyclopropyl)methanamine hydrochloride
Formel:
C₁₀H₁₂BrN·ClH Molecular Weight: 262,574 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13619675 CAS-Nummer: 1211373-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...