(1-(3-Chlorophenyl)cyclopropyl)methanamine hydrochloride

Lieferant: Apollo Scientific

OR307755-1G
APOSOR307755-1GEA 401 EUR
APOSOR307755-1G
(1-(3-Chlorophenyl)cyclopropyl)methanamine hydrochloride
(1-(3-Chlorophenyl)cyclopropyl)methanamine hydrochloride
Formel: C₁₀H₁₂ClN·ClH
Molecular Weight: 218,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864779
CAS-Nummer: 1209173-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR