(1-(3,4-Dimethoxyphenyl)cyclopropyl)methanamine hydrochloride

Lieferant: Apollo Scientific

OR307756-1G
APOSOR307756-1GEA 401 EUR
APOSOR307756-1G
(1-(3,4-Dimethoxyphenyl)cyclopropyl)methanamine hydrochloride
(1-(3,4-Dimethoxyphenyl)cyclopropyl)methanamine hydrochloride
Formel: C₁₂H₁₇NO₂·ClH
Molecular Weight: 243,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619676
CAS-Nummer: 1208550-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR