Benzo[d]isoxazole-3-carboxylic acid

Lieferant: Apollo Scientific

OR307757-1G
APOSOR307757-1GEA 128 EUR
APOSOR307757-1G
Benzo[d]isoxazole-3-carboxylic acid
Benzo[d]isoxazole-3-carboxylic acid
Formel: C₈H₅NO₃
Molecular Weight: 163.1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659786
CAS-Nummer: 28691-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR