6-Amino-2,3-dimethyl-2H-indazole

Lieferant: Apollo Scientific

Synonyme: 2,3-Dimethyl-2H-indazol-6-amine

OR307760-1G
APOSOR307760-1GEA 962 EUR
APOSOR307760-1G
6-Amino-2,3-dimethyl-2H-indazole
6-Amino-2,3-dimethyl-2H-indazole
Formel: C₉H₁₁N₃
Molecular Weight: 161,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258896
CAS-Nummer: 444731-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR