1-(3,3-Dimethyl-6-nitroindolin-1-yl)ethanone

Lieferant: Apollo Scientific

OR307761-1G
APOSOR307761-1GEA 184 EUR
APOSOR307761-1G
1-(3,3-Dimethyl-6-nitroindolin-1-yl)ethanone
1-(3,3-Dimethyl-6-nitroindolin-1-yl)ethanone
Formel: C₁₂H₁₄N₂O₃
Molecular Weight: 234.251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07636645
CAS-Nummer: 453562-68-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR