1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone hydrochloride

Lieferant: Apollo Scientific

OR307762-250MG
APOSOR307762-250MGEA 227 EUR
APOSOR307762-250MG
1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone hydrochloride
1-(6-Amino-3,3-dimethylindolin-1-yl)ethanone hydrochloride
Formel: C₁₂H₁₆N₂O·ClH
Molecular Weight: 240,729 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15530198
CAS-Nummer: 1226815-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR