4-(Pyrrolidin-1-ylmethyl)aniline hydrochloride

Lieferant: Apollo Scientific

OR307768-1G
APOSOR307768-1GEA 301 EUR
APOSOR307768-1G
4-(Pyrrolidin-1-ylmethyl)aniline hydrochloride
4-(Pyrrolidin-1-ylmethyl)aniline hydrochloride
Formel: C₁₁H₁₆N₂·ClH
Molecular Weight: 212,719 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08447180
CAS-Nummer: 866956-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR