4-((Diethylamino)methyl)aniline hydrochloride

Lieferant: Apollo Scientific

OR307769-1G
APOSOR307769-1GEA 433 EUR
APOSOR307769-1G
4-((Diethylamino)methyl)aniline hydrochloride
4-((Diethylamino)methyl)aniline hydrochloride
Formel: C₁₁H₁₈N₂·ClH
Molecular Weight: 214,735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870782
CAS-Nummer: 1204811-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR