4-(2-(Pyrrolidin-1-yl)ethyl)aniline hydrochloride

Lieferant: Apollo Scientific

OR307770-1G
APOSOR307770-1GEA 433 EUR
APOSOR307770-1G
4-(2-(Pyrrolidin-1-yl)ethyl)aniline hydrochloride
4-(2-(Pyrrolidin-1-yl)ethyl)aniline hydrochloride
Formel: C₁₂H₁₈N₂·ClH
Molecular Weight: 226,746 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870783
CAS-Nummer: 1135228-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR